Få mere at vide
Triamcinolone Acetonide-d6, CDN
Artikelnummer. 30313108
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313108 0.01 g
30319923 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313108 Leverandør CDN Leverandørnr. CDND62030.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triamcinolone Acetonide D6 (acetonide-D6)
CAS 352431-33-5
Molekylær formel C24 D6 H25 F O6
Mængde 0.01 g
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11,21-dihydroxy-16,17-[[1-(methyl-d3)ethylidene-2,2,2-d3]bis(oxy)]-, (11β,16α)- (9 CI), Triamcinolone acetonide-d6, Triamcinolone acetonide D6 (acetonide-D6), 9-Fluoro-11β,21-dihydroxy-16α,17-[1-(methyl-d3)-2,2,2-d3-ethylidenedioxy)pregna-1,4-diene-3,20-dione, 9α-Fluoro-11β,21-dihydroxy-16α,17α-(isopropylidene-d6)dioxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-hydroxyprednisolone 16α,17α-acetonide-d6, Triamcinolone acetonide D6 (acetonide D6)
InChI formel InChI=1 S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28 H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])C1(O[C@@H]2 C[C@H]3 [C@@H]4 CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO)C([2 H])([2 H])[2 H]
Molekylvægt (g/mol) 440.535
Formel vægt 440.248 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity