Få mere at vide
Triamcinolone 21-Acetate, TRC
Artikelnummer. 30452277
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30452277 25 mg -
30455982 10 mg -
30455503 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30452277 Leverandør Toronto Research Chemicals Leverandørnr. TRCT76729025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3859-65-2
Molekylær formel C23H29FO7
Molekylvægt (g/mol) 436.47
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,16α,17,21-tetrahydroxy-, 21-acetate (6CI,7CI,8CI), 9-Fluoro-11β,16α,17,21-trihydroxypregna-1,4-diene-3,20-dione, 21-acetate, Triamcinolone 21-Acetate
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16R,17S)-9-fluoro-11,16,17-trihydroxy-10,13-dimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMIL CC(=O)OCC(=O)[C@@]1(O)[C@H](O)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale Triamcinolone 21-Acetate
Mængde 25 mg
InChI formel InChI=1S/C23H29FO7/c1-12(25)31-11-19(29)23(30)17(27)9-16-15-5-4-13-8-14(26)6-7-20(13,2)22(15,24)18(28)10-21(16,23)3/h6-8,15-18,27-28,30H,4-5,9-11H2,1-3H3/t15-,16-,17+,18-,20-,21-,22-,23-/m0/s1
Formel vægt 436.1897