Få mere at vide
Triamcinolone 16,21-Diacetate, TRC
Artikelnummer. 30452373
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452373 25 mg
30452232 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30452373 Leverandør Toronto Research Chemicals Leverandørnr. TRCT76717025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67-78-7
Molekylær formel C25 H31 F O8
Molekylvægt (g/mol) 478.51
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,16α,17,21-tetrahydroxy-, 16,21-diacetate (6CI,7CI,8CI), (11β,16α)-16,21-Bis(acetyloxy)-9-fluoro-11,17-dihydroxypregna-1,4-diene-3,20-dione, 16α,21-Diacetoxy-9α-fluoro-11β,17α-dihydroxy-1,4-pregnadiene-3,20-dione, 16α-Hydroxy-9α-fluoroprednisolone diacetate, 8357RP, 9-Fluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione 16,21-diacetate, 9α-Fluoro-1,4-pregnadiene-11β,16α,17α,21-tetraol-3,20-dione 16,21-diacetate, 9α-Fluoro-16α-hydroxyprednisolone 16,21-diacetate, Aristocort Forte, Aristocort Forte Parenteral, Aristocort Parenterals, Aristocort Syrup, Aristocort diacetate forte, Aristocort diacetate parenteral, CINO 40, Cenacort, Kenacort diacetate Syrup, Kenacourt, Orion, Polcortolon, TAC-D, Tedarol, Tracilon, Triamcinolone 16,21-diacetate, Triamcinolone diacetate, Triamolone 40, δ1-16α-Hydroxy-9α-fluorohydrocortisone diacetate, δ1-9α-Fluoro-16α-hydroxyhydrocortisone diacetate
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16R,17S)-16-acetyloxy-9-fluoro-11,17-dihydroxy-10,13-dimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMIL CC(=O)OCC(=O)[C@@]1(O)[C@@H](C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)OC(=O)C
Tekniske data
Kemisk navn eller materiale Triamcinolone 16,21-Diacetate
Mængde 25 mg
InChI formel InChI=1S/C25H31FO8/c1-13(27)33-12-20(31)25(32)21(34-14(2)28)10-18-17-6-5-15-9-16(29)7-8-22(15,3)24(17,26)19(30)11-23(18,25)4/h7-9,17-19,21,30,32H,5-6,10-12H2,1-4H3/t17-,18-,19-,21+,22-,23-,24-,25+/m0/s1
Formel vægt 478.2003