Få mere at vide
Triamcinolone 16,21-Diacetate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Triamcinolone 16,21-Diacetate |
| CAS | 67-78-7 |
| Analyt- eller komponentnavne | Triamcinolone 16,21-Diacetate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H31 F O8 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Triamcinolone Imp. A (EP), Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,16alpha,17,21-tetrahydroxy-, 16,21-diacetate (6CI,7CI,8CI), (11beta,16alpha)-16,21-Bis(acetyloxy)-9-fluoro-11,17-dihydroxypregna-1,4-diene-3,20-dione, 16alpha,21-Diacetoxy-9alpha-fluoro-11beta,17alpha-dihydroxy-1,4-pregnadiene-3,20-dione, 16alpha-Hydroxy-9alpha-fluoroprednisolone diacetate, 8357RP, 9-Fluoro-11beta,16alpha,17,21-tetrahydroxypregna-1,4-diene-3,20-dione 16,21-diacetate, 9alpha-Fluoro-1,4-pregnadiene-11beta,16alpha,17alpha,21-tetraol-3,20-dione 16,21-diacetate, 9alpha-Fluoro-16alpha-hydroxyprednisolone 16,21-diacetate, Aristocort Forte, Aristocort Forte Parenteral, Aristocort Parenterals, Aristocort Syrup, Aristocort diacetate forte, Aristocort diacetate parenteral, CINO 40, Cenacort, Kenacort diacetate Syrup, Kenacourt, Orion, Polcortolon, TAC-D, Tedarol, Tracilon, Triamcinolone 16,21-diacetate, Triamcinolone diacetate, Triamolone 40, Delta1-16alpha-Hydroxy-9alpha-fluorohydrocortisone diacetate, Delta1-9alpha-Fluoro-16alpha-hydroxyhydrocortisone diacetate, Triamcinolone Impurity A |
| InChI formel | InChI=1S/C25H31FO8/c1-13(27)33-12-20(31)25(32)21(34-14(2)28)10-18-17-6-5-15-9-16(29)7-8-22(15,3)24(17,26)19(30)11-23(18,25)4/h7-9,17-19,21,30,32H,5-6,10-12H2,1-4H3/t17-,18-,19-,21+,22-,23-,24-,25+/m0/s1 |
| Løsningstype | Neat |
| SMIL | CC(=O)OCC(=O)[C@@]1(O)[C@@H](C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)OC(=O)C |
| Vis mere |