Få mere at vide
Triadimenol-d4 (4-chlorophenoxy-d4) (mixture of stereoisomers), CDN
Artikelnummer. 30309797
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309797 0.005 g
30319309 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309797 Leverandør CDN Leverandørnr. CDND76840.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triadimenol D4 (Phenoxy D4)
CAS 2121989-56-6
Molekylær formel C14 D4 H14 Cl N3 O2
Mængde 0.005 g
Synonym Triadimenol D4 (Phenoxy D4)
InChI formel InChI=1 S/C14H18ClN3O2/c1-14(2,3)12(19)13(18-9-16-8-17-18)20-11-6-4-10(15)5-7-11/h4-9,12-13,19 H,1-3H3/i4D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c(OC(C(O)C(C)(C)C)n2cncn2)c([2 H])c([2 H])c1Cl
IUPAC navn 1-(4-chloro-2,3,5,6-tetradeuterio-phenoxy)-3,3-dimethyl-1-(1,2,4-triazol-1-yl)butan-2-ol
Molekylvægt (g/mol) 299.79
Formel vægt 299.1339 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity