Få mere at vide
Triacetin-d9, TRC
Artikelnummer. 30452514
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30452514 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452514 Leverandør Toronto Research Chemicals Leverandørnr. TRCT720853500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triacetin-d9
CAS 2733570-86-8
Molekylær formel C9 D9 H5 O6
Mængde 500 mg
Synonym 1,2,3-Propanetriol tri(acetate-d3), 1,2,3-Propanetriol triacetate-d9, Triacetin-d9, Glycerol tri(acetate-d3), Tri(acetyl-d3)glycerol, Triacetin-D9 (triacetyl-D9)
InChI formel InChI=1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(=O)OCC(COC(=O)C([2H])([2H])[2H])OC(=O)C([2H])([2H])[2H]
IUPAC navn 2,3-bis[(2,2,2-trideuterioacetyl)oxy]propyl 2,2,2-trideuterioacetate
Molekylvægt (g/mol) 227.2593
Formel vægt 227.1355