Få mere at vide
Triacetin-d5, TRC
Artikelnummer. 30452921
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30452921 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452921 Leverandør Toronto Research Chemicals Leverandørnr. TRCT72085250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triacetin-d5
CAS 159510-46-0
Molekylær formel C9 D5 H9 O6
Mængde 50 mg
Synonym 1,2,3-Propane-1,1,2,3,3-d5-triol','triacetate (9CI), 1,2,3-Propane-1,1,2,3,3-d5-triol triacetate, 1,2,3-Propanetriol-1,1,2,3,3-d5 triacetate, Triacetin-d5, Glycerol-d5 triacetate, Triacetylglycerol-d5, Triacetin-D5 (glycerol-D5)
InChI formel InChI=1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3/i4D2,5D2,9D
SMIL [2H]C([2H])(OC(=O)C)C([2H])(OC(=O)C)C([2H])([2H])OC(=O)C
IUPAC navn (2,3-diacetyloxy-1,1,2,3,3-pentadeuteriopropyl) acetate
Molekylvægt (g/mol) 223.2347
Formel vægt 223.1104