missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
tri-Sodium Phosphate Dodecahydrate, Supelco™
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tri-Sodium Phosphate Dodecahydrate |
| Smeltepunkt | 75°C (Elimination of water of crystallisation) |
| Mængde | 5 kg |
| MDL nummer | MFCD00149198 |
| Synonym | tri-Sodium phosphate dodecahydrate, Sodium phosphate tribasic, Trisodium phosphate, tri-Sodium phosphate dodecahydrate |
| Molekylvægt (g/mol) | 380.12 |
| Grad | Research |
| Tæthed | 1.62 g/cm3 at 20°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?