Få mere at vide
Tri-p-tolylamine-d21, CDN
Artikelnummer. 30316416
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316416 0.05 g -
30322160 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316416 Leverandør CDN Leverandørnr. CDND22090.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tri-p-tolylamine-D21
CAS 201944-90-3
Molekylær formel C21 D21 N
Mængde 0.05 g
Synonym Benzen-2,3,5,6-d4-amine, 4-(methyl-d3)-N,N-bis[4-(methyl-d3)phenyl-2,3,5,6-d4]- (9 CI), Tri-p-tolylamine-d21, Tri-p-tolylamine-D21, N,N,N-Tri[4-(methyl-d3)phenyl-d4]amine, N,N,N-Tri(4-tolyl-d7)amine, Tris(4-methylphenyl)amine-d21, 4-(Methyl-d3)-N,N-bis[4-(methyl-d3)phenyl-d4]benzenamine
InChI formel InChI=1 S/C21H21N/c1-16-4-10-19(11-5-16)22(20-12-6-17(2)7-13-20)21-14-8-18(3)9-15-21/h4-15 H,1-3H3/i1D3,2D3,3D3,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D,15 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N(c2c([2 H])c([2 H])c(c([2 H])c2[2 H])C([2 H])([2 H])[2 H])c3c([2 H])c([2 H])c(c([2 H])c3[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,3,5,6-tetradeuterio-N,N-bis[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenyl]-4-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 308.527
Formel vægt 308.299 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity