Få mere at vide
Tri(n-octyl-1,1-d2)amine, CDN
Artikelnummer. 30309817
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30309817 0.05 g -
30318986 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309817 Leverandør CDN Leverandørnr. CDND58300.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trioctylamine-1,1,1',1',1'',1''-D6
CAS 1219798-90-9
Molekylær formel C24 D6 H45 N
Mængde 0.05 g
Synonym Tri(n-octyl-1,1-d2)amine, Tri(octyl-1,1-d2)amine, N,N-Di(octyl-1,1-d2)-1-octanamine-1,1-d2, Trioctylamine-d6, N,N,N-Trioctylamine-d6, Trioctylamine-1,1,1',1',1'',1''-D6, Trioctylamine-1,1,1',1',1'',1''-D6
InChI formel InChI=1 S/C24H51N/c1-4-7-10-13-16-19-22-25(23-20-17-14-11-8-5-2)24-21-18-15-12-9-6-3/h4-24H2,1-3H3/i22D2,23D2,24D2
SMIL [2 H]C([2 H])(CCCCCCC)N(C([2 H])([2 H])CCCCCCC)C([2 H])([2 H])CCCCCCC
IUPAC navn 1,1-dideuterio-N,N-bis(1,1-dideuteriooctyl)octan-1-amine
Molekylvægt (g/mol) 359.705
Formel vægt 359.44 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity