missing translation for 'onlineSavingsMsg'
Få mere at vide

Tri-n-hexyl Trimellitate, TRC

Artikelnummer. 30417411
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417411 50 mg
30413718 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30417411 Leverandør Toronto Research Chemicals Leverandørnr. TRCT26395050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Tri-n-hexyl Trimellitate
CAS 1528-49-0
Molekylær formel C27H42O6
Mængde 50 mg
Synonym trihexyl benzene-1,2,4-tricarboxylate
InChI formel InChI=1S/C27H42O6/c1-4-7-10-13-18-31-25(28)22-16-17-23(26(29)32-19-14-11-8-5-2)24(21-22)27(30)33-20-15-12-9-6-3/h16-17,21H,4-15,18-20H2,1-3H3
SMIL O=C(OCCCCCC)C1=CC=C(C(=O)OCCCCCC)C(=C1)C(=O)OCCCCCC
IUPAC navn trihexyl benzene-1,2,4-tricarboxylate
Molekylvægt (g/mol) 462.62
Formel vægt 462.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.