Learn More
Alfa Aesar™ Tri-n-butyltin trifluoromethanesulfonate, tech. 90%
Brand: Alfa Aesar™ H55192.03
Additional Details : CAS Number : 68725-14-4 Weight : 0.00100kg Transport : UN number : 2923 Chem class : 8 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
68725-14-4 | |
MFCD00043324 | |
Tri-n-butyltin triflate |
Specifications
1g | |
Moisture Sensitive | |
68725-14-4 | |
MFCD00043324 | |
5818090 | |
439.1 | |
90% |
Tri-n-butyltin trifluoromethanesulfonate | |
Brown to White | |
CF3SO3Sn[(CH2)3CH3]3 | |
UN2923 | |
Tri-n-butyltin triflate | |
Solid | |
Technical |