Få mere at vide
Tri-n-butyl 2-Acetyl-d3-citrate, CDN
Artikelnummer. 30321882
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321882 0.05 g -
30320138 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321882 Leverandør CDN Leverandørnr. CDND79260.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tributyl O-Acetylcitrate-d3
CAS 1794753-49-3
Molekylær formel C20D3H31O8
Mængde 0.05 g
Synonym 2-(Acetyloxy-d3)-1,2,3-propanetricarboxylic Acid 1,2,3-Tributyl Ester, Citric Acid Tributyl Ester Acetate-d3, ATBC-d3, Acetyl-d3 Butyl Citrate, Acetyl-d3 Tributyl Citrate, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3, Citroflex 4 A-S-d3
InChI formel InChI=1 S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3/i4D3
SMIL [2 H]C([2 H])([2 H])C(=O)OC(CC(=O)OCCCC)(CC(=O)OCCCC)C(=O)OCCCC
IUPAC navn tributyl 2-(2,2,2-trideuterioacetyl)oxypropane-1,2,3-tricarboxylate
Molekylvægt (g/mol) 405.5
Formel vægt 405.2442 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity