missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TREM1 Antibody (L5-B8.2A12.3A12), Alexa Fluor™ 350 , Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-40907AF350
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TREM1 Monoclonal antibody specifically detects TREM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Tekniske data
| TREM1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Mouse TREM-1 Fc fusion protein. | |
| 0.25 mL | |
| Signal Transduction | |
| 54210 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| L5-B8.2A12.3A12 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion