missing translation for 'onlineSavingsMsg'
Få mere at vide

Trazodone Isobutyl Ether, TRC

Artikelnummer. 30414775
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414775 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30414775 Leverandør Toronto Research Chemicals Leverandørnr. TRCT242465250MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1-(3-chlorophenyl)-4-[3-(2-methylpropoxy)propyl]piperazine
CAS 2470436-00-9
Molekylær formel C17 H27 Cl N2 O
Mængde 250 mg
Synonym Trazodone Isobutyl Ether Analog
InChI formel InChI=1S/C17H27ClN2O/c1-15(2)14-21-12-4-7-19-8-10-20(11-9-19)17-6-3-5-16(18)13-17/h3,5-6,13,15H,4,7-12,14H2,1-2H3
SMIL CC(C)COCCCN1CCN(CC1)c2cccc(Cl)c2
IUPAC navn 1-(3-chlorophenyl)-4-[3-(2-methylpropoxy)propyl]piperazine
Molekylvægt (g/mol) 310.862
Formel vægt 310.181
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.