Få mere at vide
Trazodone EP Impurity F (as Hydrochloride), Mikromol™
Beskrivelse
Trazodone EP Impurity F (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride |
| CAS | 52605-52-4 |
| Analyt- eller komponentnavne | 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C13 H18 Cl2 N2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride, Trazodone Hydrochloride Imp. F (BP) as Hydrochloride, Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, hydrochloride (1:1), Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, monohydrochloride (9CI), 1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride (1:1), Nefazodone Related Compound A as Hydrochloride, Trazodone Related Compound F as Hydrochloride, Trazodone Hydrochloride Impurity F as Hydrochloride, Trazodone Impurity F as Hydrochloride |
| InChI formel | InChI=1S/C13H18Cl2N2.ClH/c14-5-2-6-16-7-9-17(10-8-16)13-4-1-3-12(15)11-13;/h1,3-4,11H,2,5-10H2;1H |
| SMIL | Cl.ClCCCN1CCN(CC1)c2cccc(Cl)c2 |
| IUPAC navn | 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazine;hydrochloride |
| Vis mere |