Få mere at vide
Trazodone EP Impurity F (as Hydrochloride), Mikromol™
Artikelnummer. 30050133
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050133 100 mg
30052224 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050133 Leverandør Mikromol Leverandørnr. MM0421.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trazodone EP Impurity F (as Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride
CAS 52605-52-4
Analyt- eller komponentnavne 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C13 H18 Cl2 N2 . Cl H
Certificeringer/compliance ISO 9001
Synonym 1-(3-Chloropropyl)-4-(3-chlorophenyl)piperazine Hydrochloride, Trazodone Hydrochloride Imp. F (BP) as Hydrochloride, Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, hydrochloride (1:1), Piperazine, 1-(3-chlorophenyl)-4-(3-chloropropyl)-, monohydrochloride (9CI), 1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride (1:1), Nefazodone Related Compound A as Hydrochloride, Trazodone Related Compound F as Hydrochloride, Trazodone Hydrochloride Impurity F as Hydrochloride, Trazodone Impurity F as Hydrochloride
InChI formel InChI=1S/C13H18Cl2N2.ClH/c14-5-2-6-16-7-9-17(10-8-16)13-4-1-3-12(15)11-13;/h1,3-4,11H,2,5-10H2;1H
SMIL Cl.ClCCCN1CCN(CC1)c2cccc(Cl)c2
IUPAC navn 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazine;hydrochloride
Molekylvægt (g/mol) 309.66
Mængde 100 mg
Formel vægt 308.0614 g/mol