Få mere at vide
Trazodone-d6 Hydrochloride, TRC
Artikelnummer. 30456419
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30456419 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456419 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7185021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trazodone-D6 Hydrochloride
CAS 1181578-71-1
Molekylær formel C19 2H6 H16 Cl N5 O . Cl H
Mængde 1 mg
Synonym Trazodone-D6 Hydrochloride
InChI formel InChI=1S/C19H22ClN5O.ClH/c20-16-5-3-6-17(15-16)23-13-11-22(12-14-23)8-4-10-25-19(26)24-9-2-1-7-18(24)21-25;/h1-3,5-7,9,15H,4,8,10-14H2;1H/i4D2,8D2,10D2;
SMIL Cl.[2H]C([2H])(N1CCN(CC1)c2cccc(Cl)c2)C([2H])([2H])C([2H])([2H])N3N=C4C=CC=CN4C3=O
IUPAC navn 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]-1,1,2,2,3,3-hexadeuteriopropyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
Molekylvægt (g/mol) 414.36
Formel vægt 413.1656