missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Travoprost, MedChemExpress

Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Description
Travoprost (Fluprostenol isopropyl ester), an isopropyl ester prodrug, is a high affinity, selective FP prostaglandin full receptor agonist. Travoprost has the ocular hypotensive efficacy and has the potential for glaucoma and ocular hypertension.
Specifications
Specifications
Chemical Name or Material | Travoprost |
CAS | 157283-68-6 |
Color | Colorless |
Purity Grade Notes | Research |
Molecular Formula | C26H35F3O6 |
Quantity | 1 mg |
Synonym | Fluprostenol isopropyl ester AL6221 Flu-Ipr |
Solubility Information | Ethanol : 60 mg/mL (119.87 mM; Need ultrasonic) ∣DMSO : ≥ 41.67 mg/mL (83.25 mM) |
SMILES | O=C(OC(C)C)CCC/C=C\C[C@@H]1[C@@H](/C=C/[C@@H](O)COC2=CC=CC(C(F)(F)F)=C2)[C@H](O)C[C@@H]1O |
Molecular Weight (g/mol) | 500.55 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Travoprost, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction