Få mere at vide
Traumatic Acid, TRC
Artikelnummer. 30455902
Skift visning
Mængde:
100 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455902 100 mg
30452256 500 mg
30450737 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30455902 Leverandør Toronto Research Chemicals Leverandørnr. TRCT715500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6402-36-4
Molekylær formel C12H20O4
Molekylvægt (g/mol) 228.28
Synonym (E)-2-Dodecenedioic Acid, trans-2-Dodecenedioic Acid, trans-Traumatic Acid
IUPAC navn (E)-dodec-2-enedioic acid
SMIL OC(=O)CCCCCCCC\C=C\C(=O)O
Tekniske data
Kemisk navn eller materiale Traumatic Acid
Mængde 100 mg
InChI formel InChI=1S/C12H20O4/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16/h7,9H,1-6,8,10H2,(H,13,14)(H,15,16)/b9-7+
Formel vægt 228.1362