Få mere at vide
Tranylcypromine USP Related Compound A (as Hydrochloride), Mikromol™
Artikelnummer. 30050479
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050479 25 mg
30051616 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30050479 Leverandør Mikromol Leverandørnr. MM1687.020025
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tranylcypromine USP Related Compound A (as Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac cis-2-Phenylcyclopropylamine Hydrochloride
CAS 54779-58-7
Analyt- eller komponentnavne rac cis-2-Phenylcyclopropylamine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C9 H11 N . Cl H
Certificeringer/compliance ISO 9001
Synonym rac-cis-2-Phenylcyclopropanamine Hydrochloride (cis-Tranylcypromine Hydrochloride), Cyclopropanamine, 2-phenyl-, hydrochloride, (1R,2R)-rel-, Cyclopropanamine, 2-phenyl-, hydrochloride, cis-, cis-Tranylcypromine hydrochloride, rac-cis-2-Phenylcyclopropanamine Hydrochloride, Tranylcypromine Related Compound A as Hydrochloride
InChI formel InChI=1S/C9H11N.ClH/c10-9-6-8(9)7-4-2-1-3-5-7;/h1-5,8-9H,6,10H2;1H/t8-,9-;/m1./s1
SMIL Cl.N[C@@H]1C[C@@H]1c2ccccc2
IUPAC navn (1R,2R)-2-phenylcyclopropan-1-amine;hydrochloride
Molekylvægt (g/mol) 169.65
Mængde 25 mg
Formel vægt 169.0658 g/mol