Få mere at vide
Tranylcypromine Sulfate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tranylcypromine Sulfate |
| CAS | 13492-01-8 |
| Analyt- eller komponentnavne | Tranylcypromine Sulfate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | 2 C9 H11 N . H2 O4 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Cyclopropanamine, 2-phenyl-, (1R,2S)-rel-, sulfate (2:1), Cyclopropanamine, 2-phenyl-, trans-, sulfate (2:1), Parnate, Parnetil, Tranylcypromine sulfate, Tylciprine, dl-Tranylcypromine sulfate, trans-2-Phenylcyclopropanamine sulfate (2:1), Cyclopropanamine, 2-phenyl-, trans-(+/-)-, sulfate (2:1), Cyclopropylamine, 2-phenyl-, sulfate (2:1), trans-(+/-)- (8CI) |
| InChI formel | InChI=1S/2C9H11N.H2O4S/c2*10-9-6-8(9)7-4-2-1-3-5-7;1-5(2,3)4/h2*1-5,8-9H,6,10H2;(H2,1,2,3,4)/t2*8-,9+;/m00./s1 |
| Løsningstype | Neat |
| SMIL | N[C@@H]1C[C@H]1c2ccccc2.N[C@@H]3C[C@H]3c4ccccc4.OS(=O)(=O)O |
| Vis mere |