missing translation for 'onlineSavingsMsg'
Få mere at vide

Tranylcypromine hemisulfate, MedChemExpress

Artikelnummer. 16478776
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16478776 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16478776 Leverandør Medchem Express Leverandørnr. HYB1496

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Tranylcypromine hemisulfate (dl-Tranylcypromine hemisulfate) is an irreversible, nonselective monoamine oxidase (MAO) inhibitor used in the treatment of depression. Tranylcypromine hemisulfate is also a lysine-specific demethylase 1 (LSD1) inhibitor, suppresses lesion growth and improves generalized hyperalgesia in mouse with induced endometriosis. Tranylcypromine has antidepressant effects.

Tranylcypromine hemisulfate (dl-Tranylcypromine hemisulfate) is an irreversible, nonselective monoamine oxidase (MAO) inhibitor used in the treatment of depression. Tranylcypromine hemisulfate is also a lysine-specific demethylase 1 (LSD1) inhibitor, suppresses lesion growth and improves generalized hyperalgesia in mouse with induced endometriosis. Tranylcypromine has antidepressant effects.

Tekniske data

Kemisk navn eller materiale Tranylcypromine hemisulfate
CAS 13492-01-8
Farve White
Renhedsgrad noter Research
Molekylær formel C9H12NO2S0.5
Mængde 100 mg
Synonym dl-Tranylcypromine hemisulfate trans-2-Phenylcyclopropylamine hemisulfate salt
Opløselighedsinformation H2O : 25 mg/mL (137.19 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (27.44 mM; ultrasonic and warming and heat to 60°C)
SMIL N[C@H]1[C@H](C2=CC=CC=C2)C1.[0.5H2SO4]
Molekylvægt (g/mol) 182.23
Formel vægt 182.23
Procent renhed 99.28%
Grad Research
Fysisk form Powder
Til brug med (applikation) Neuroscience-Neurodegeneration
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.