Få mere at vide
Kartell™ Transparent Polypropylene Powder Funnels
BIOPHARMA
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
Brand:  Kartell™ 0016800
Produktkode 12117790
  • 523.00 DKK / Pakke med 20
Kun null tilbage
Forventet forsendelse: 19-01-2026
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
  • 60° angled sides
  • External ribbing avoids pressure lock
  • Can be used with liquids
  • Autoclavable