Learn More
Fisherbrand™ Translucent Silicone Tubing
Product Code. 11756599 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer:
9 mm
12 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
11756599 12 mm Each
11746599 9 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11756599
  • ONLINE EXCLUSIVE PRICE
    232.17 DKK
    168.82 DKK / Pakke med 1
    Spare 63.35 DKK
    27% Slukket
Forventet forsendelse: 07-12-2026
Providing high tear strength tubing for the laboratory. Fisherbrand™ Translucent Silicone Tubing is available in a comprehensive range of sizes.
  • Autoclavable
  • Non-yellowing
  • High gloss finish
  • Excellent quality
  • Extended life in Peristaltic pumps
  • Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Autoclavable Autoclavable
Diameter (Metric) Inner 8 mm
Material Silicone
Diameter (Metric) Outer 12 mm
Overall Length (Metric) 4 m