Learn More
Ratiolab™ Pasteur-Plast Makro Pipets
Product Code. 15253837
Change view
Click to view available options
Volume (Metric):
3 mL
Unit Size:
Pack of 500
Product Code. Volume (Metric) Unit Size
15253837 3 mL Pack of 500
1 options
Product Code. 15253837 Supplier Ratiolab™ Supplier No. 2600111
  • ONLINE EXCLUSIVE PRICE
    193.00 DKK
    87.93 DKK / Pakke med 500
    Spare 105.07 DKK
    54% Slukket
Forventet forsendelse: 15-12-2026
Use these pipettes for simple and safe liquid handling. ratiolab™ Pasteur-Plast Makro Pipets are inert to biological fluids and most acids. The low affinity of the surface reduces the loss of cells and valuable proteins through binding.
Cross-contamination and carry-over simply cannot occur so that tedious clean-ups are unnecessary. In addition, there is no danger of glass breakage with the resulting danger of infectious material escaping.
  • New soft, elastic bulb
  • Unbreakable – thus reduced risk of glass injury
  • Burr-free orifice for neat, clean and precise dispensing
  • Embossed, external graduations
  • BSE-free can be used for food
Specifications
Sterility Non-sterile
Quantity 500 Pack
Volume (Metric) 3 mL
Type Pasteur Pipet