Få mere at vide
trans,trans-Muconic-d4 Acid, CDN
Artikelnummer. 30319737
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319737 0.01 g
30309798 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319737 Leverandør CDN Leverandørnr. CDND62570.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans,trans-Muconic Acid D4
CAS 1955496-83-9
Molekylær formel C6 D4 H2 O4
Mængde 0.01 g
Synonym (2 E,4 E)-2,4-Hexadienedioic-2,3,4,5-d4 acid, (E,E)-Muconic acid-d4, trans,trans-1,2,3,4-Tetradeuterio-1,3-butadiene-1,4-dicarboxylic acid, trans,trans-Muconic-d4 Acid
InChI formel InChI=1 S/C6H6O4/c7-5(8)3-1-2-4-6(9)10/h1-4 H,(H,7,8)(H,9,10)/b3-1+,4-2+/i1D,2 D,3 D,4 D
SMIL [2 H]\C(=C(\[2 H])/C(=O)O)\C(=C(/[2 H])\C(=O)O)\[2 H]
IUPAC navn (2 E,4 E)-2,3,4,5-tetradeuteriohexa-2,4-dienedioic acid
Molekylvægt (g/mol) 146.1341
Formel vægt 146.0517 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity