Få mere at vide
trans,trans-Muconic Acid-d4, TRC
Artikelnummer. 30476818
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30476818 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476818 Leverandør Toronto Research Chemicals Leverandørnr. TRCM7910005MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans,trans-Muconic Acid-d4
CAS 1185239-59-1
Renhedsgrad noter HPLC
Molekylær formel C6 H2 D4 O4
Mængde 5 mg
Synonym Muconic acid-d4','trans,trans-Muconic Acid-d4','trans-trans-Muconic acid-d4','trans,trans-Muconic-d4 Acid,
InChI formel InChI=1S/C6H6O4/c7-5(8)3-1-2-4-6(9)10/h1-4H,(H,7,8)(H,9,10)/b3-1+,4-2+/i1D,2D,3D,4D
SMIL [2H]/C(C(O)=O)=C(\C([2H])=C(C(O)=O)/[2H])[2H]
IUPAC navn (2E,4E)-hexa-2,4-dienedioic-d4 acid
Molekylvægt (g/mol) 146.13
Formel vægt 146.05
Procent renhed >95