Få mere at vide
trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid, 97%, Thermo Scientific™
Artikelnummer. 15422006
Skift visning
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15422006 25 g 25g
15412006 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15422006 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H55271.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 65355-32-0
Molekylær formel C16H28O2
Molekylvægt (g/mol) 252.398
MDL nummer MFCD05864407
InChI nøgle JXPGQFKJNKWDKP-UHFFFAOYSA-N
Synonym trans-4'-propyl-1,1'-bicyclohexyl-4-carboxylic acid, 1,1'-bicyclohexyl-4-carboxylic acid, 4'-propyl, 4-4-propylcyclohexyl cyclohexanecarboxylic acid, 4-4-propylcyclohexyl cyclohexane-1-carboxylic acid, trans-4'-propyl-1,1'-bicyclohexyl-4-carboxylicacid, jxpgqfkjnkwdkp-ktslabgisa-n, 4'-propylbicyclohexane-4-carboxylic acid, 4-propyl-4'-bicyclohexanecarboxylic acid, trans-4-propylbis cyclohexane carboxylic acid, 4'-propyl-1,1'-bicyclohexyl-4-carboxylic acid
PubChem CID 793726
IUPAC navn 4-(4-propylcyclohexyl)cyclohexane-1-carboxylic acid
SMIL CCCC1CCC(CC1)C2CCC(CC2)C(=O)O
Tekniske data
Kemisk navn eller materiale trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid
Smeltepunkt 200°C
Mængde 25 g
Formel vægt 252.4
Procent renhed 97%