Få mere at vide
trans-trans-10,11-Epoxy Farnesenic Acid-d3 Methyl Ester, TRC
Artikelnummer. 30371302
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30371302 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371302 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5894022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-trans-10,11-Epoxy Farnesenic Acid-d3 Methyl Ester
CAS 951116-89-5
Renhedsgrad noter HPLC
Molekylær formel C16H23D3O3
Mængde 2.5 mg
Synonym (2E,6E)- 9-(3,3-Dimethyl-2-oxiranyl)-3,7-dimethyl-2,6-nonadienoic Acid Methyl-d3 Ester, (E,E)-(±)-10,11-Epoxy-3,7,11-trimethyl-2,6-dodecadienoic Acid Methyl-d3 Ester, (E,E)-Methyl-d3 3,7,11-trimethyl-10,11-epoxy-2,6-dodecadienoate, (±)-Juvenile Hormone III-d3, Methyl-d3 RS-(±)-10,11-Epoxyfarnesate-d3
InChI formel InChI=1S/C16H26O3/c1-12(9-10-14-16(3,4)19-14)7-6-8-13(2)11-15(17)18-5/h7,11,14H,6,8-10H2,1-5H3/b12-7+,13-11+/i5D3
SMIL O=C(OC([2H])([2H])[2H])/C=C(C)/CC/C=C(C)/CCC1OC1(C)C
Molekylvægt (g/mol) 269.39
Formel vægt 269.207
Procent renhed >95