Få mere at vide
trans-Stilbene-alpha,beta-13C2, CDN
Artikelnummer. 30319673
Skift visning
Mængde:
0.25 g
Artikelnummer. Mængde
30319673 0.25 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319673 Leverandør CDN Leverandørnr. CDNC30260.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-Stilbene-alpha,beta-13C2
CAS 153610-50-5
Molekylær formel 13C2 C12 H12
Mængde 0.25 g
Synonym Benzene, 1,1'-[(1 E)-1,2-ethenediyl-1,2-13C2]bis- (9 CI), Benzene, 1,1'-(1,2-ethenediyl-1,2-13C2)bis-, (E)- (ZCI), 1,1'-[(1 E)-1,2-ethenediyl-1,2-13C2]bisbenzene, trans-Stilbene-13C2, trans-Stilbene-α,β-13C2, trans-Stilbene-13C2 (ethylene-13C2)
InChI formel InChI=1 S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12 H/b12-11+/i11+1,12+1
SMIL [13 CH](=[13 CH]\c1ccccc1)/c2ccccc2
IUPAC navn [(E)-2-phenyl(1,2-^{13}C_{2})ethenyl]benzene
Molekylvægt (g/mol) 182.23
Formel vægt 182.101 g/mol
Procent renhed 99 atom % 13 C, min 98% Chemical Purity