Få mere at vide
trans-Resveratrol 4’-O-Beta-D-Glucuronide, TRC
Artikelnummer. 30137942
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137942 10 mg
30006189 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137942 Leverandør Toronto Research Chemicals Leverandørnr. TRCR15002510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 387372-20-5
Molekylær formel C20 H20 O9
Molekylvægt (g/mol) 404.37
Synonym Trans-Resveratrol 4'-O-B-D-Glucuronide
IUPAC navn (2S,3S,4S,5R,6S)-6-[4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]phenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL O[C@@H]1[C@@H](O)[C@H](Oc2ccc(\C=C\c3cc(O)cc(O)c3)cc2)O[C@@H]([C@H]1O)C(=O)O
Tekniske data
Kemisk navn eller materiale trans Resveratrol 4'-O-beta-D-Glucuronide
Mængde 10 mg
InChI formel InChI=1S/C20H20O9/c21-12-7-11(8-13(22)9-12)2-1-10-3-5-14(6-4-10)28-20-17(25)15(23)16(24)18(29-20)19(26)27/h1-9,15-18,20-25H,(H,26,27)/b2-1+/t15-,16-,17+,18-,20+/m0/s1
Formel vægt 404.1107