Få mere at vide
trans-p-Coumaric-d6 Acid, TRC
Artikelnummer. 30353258
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30353258 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353258 Leverandør Toronto Research Chemicals Leverandørnr. TRCC75539225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-p-Coumaric Acid D6
CAS 2708298-33-1
Molekylær formel C9 D6 H2 O3
Mængde 25 mg
Synonym (2E)-3-(4-Hydroxyphenyl-2,3,5,6-d4)-2-propenoic-2,3-d2 acid, trans-p-Coumaric acid-d6, trans-4-Hydroxycinnamic-d6 acid, trans-p-Coumaric acid D6, trans-4-Hydroxycinnamic-α,β,2,3,5,6-d6 acid
InChI formel InChI=1S/C9H8O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6,10H,(H,11,12)/b6-3+/i1D,2D,3D,4D,5D,6D
SMIL [2H]\C(=C(\[2H])/c1c([2H])c([2H])c(O)c([2H])c1[2H])\C(=O)O
IUPAC navn (E)-2,3-dideuterio-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)prop-2-enoic acid
Molekylvægt (g/mol) 170.195
Formel vægt 170.085