Få mere at vide
trans-Nerolidol, TRC
Artikelnummer. 30133734
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133734 100 mg
30136097 1 g
30007210 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133734 Leverandør Toronto Research Chemicals Leverandørnr. TRCN385710100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 40716-66-3
Molekylær formel C15 H26 O
Molekylvægt (g/mol) 222.37
Synonym 1,6,10-Dodecatrien-3-ol, 3,7,11-trimethyl-, (6E)-, 1,6,10-Dodecatrien-3-ol, 3,7,11-trimethyl-, (E)-, (6E)-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, (6E)-3,7,11-Trimethyldodeca-1,6,10-trien-3-ol, (6E)-Nerolidol, (E)-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, (E)-Nerolidol, (±)-trans-Nerolidol, 1-Geranyl-2-methyl-3-buten-2-ol, Nerolidol B, trans-Nerolidol
IUPAC navn (6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
SMIL CC(=CCC\C(=C\CCC(C)(O)C=C)\C)C
Tekniske data
Kemisk navn eller materiale trans-Nerolidol
Mængde 100 mg
InChI formel InChI=1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11+
Formel vægt 222.1984