Få mere at vide
trans-Glutaconic Acid, TRC
Artikelnummer. 30240862
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240862 500 mg
30240721 5 g
30240616 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240862 Leverandør Toronto Research Chemicals Leverandørnr. TRCG596940500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 628-48-8
Molekylær formel C5H6O4
Molekylvægt (g/mol) 130.1
Synonym (E)-2-Pentenedioic Acid, (E)-Glutaconic Acid, trans-Glutaconic Acid, trans-Glutaconic Acid, (2E)-2-Pentenedioic acid
IUPAC navn (E)-pent-2-enedioic acid
SMIL OC(=O)C\C=C\C(=O)O
Tekniske data
Kemisk navn eller materiale trans-Glutaconic Acid
Mængde 500 mg
InChI formel InChI=1S/C5H6O4/c6-4(7)2-1-3-5(8)9/h1-2H,3H2,(H,6,7)(H,8,9)/b2-1+
Formel vægt 130.0266