Få mere at vide
trans-Doxercalciferol, TRC
Artikelnummer. 30134314
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134314 10 mg
30011969 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134314 Leverandør Toronto Research Chemicals Leverandørnr. TRCD55254010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 74007-20-8
Molekylær formel C28 H44 O2
Molekylvægt (g/mol) 412.65
Synonym 1,3-Cyclohexanediol, 4-methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-7a-methyl-1-[(1R,2E,4R)-1,4,5-trimethyl-2-hexen-1-yl]-4H-inden-4-ylidene]ethylidene]-, (1R,3S,5E)-, 9,10-Secoergosta-5,7,10(19),22-tetraene-1,3-diol, (1α,3β,5E,7E,22E)- (9CI), (1R,3S,5E)-4-Methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-7a-methyl-1-[(1R,2E,4R)-1,4,5-trimethyl-2-hexen-1-yl]-4H-inden-4-ylidene]ethylidene]-1,3-cyclohexanediol, trans-Doxercalciferol, (5E)-Doxercalciferol, (1α,3β,5E,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraene-1,3-diol
IUPAC navn (1R,3S,5E)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
SMIL CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C\3/C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C
Tekniske data
Kemisk navn eller materiale trans-Doxercalciferol
Mængde 10 mg
InChI formel InChI=1S/C28H44O2/c1-18(2)19(3)9-10-20(4)25-13-14-26-22(8-7-15-28(25,26)6)11-12-23-16-24(29)17-27(30)21(23)5/h9-12,18-20,24-27,29-30H,5,7-8,13-17H2,1-4,6H3/b10-9+,22-11+,23-12+/t19-,20+,24+,25+,26-,27-,28+/m0/s1
Formel vægt 412.3341