Få mere at vide
trans-Crotoxyphos, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Crotoxyphos |
| CAS | 7700-17-6 |
| Molekylær formel | C14 H19 O6 P |
| Mængde | 10 mg |
| Synonym | 2-Butenoic acid, 3-[(dimethoxyphosphinyl)oxy]-, 1-phenylethyl ester, (E)-, Crotonic acid, 3-hydroxy-, α-methylbenzyl ester, dimethyl phosphate, (E)- (8CI), 3-Hydroxycrotonic acid α-methylbenzyl ester dimethyl phosphate, Ciodrin, Crotoxyfos, Crotoxyphos, Cyodrin, Dimethyl 2-(α-methylbenzyloxycarbonyl)-1-methylvinyl phosphate, O,O-Dimethyl O-[1-methyl-2-(1-phenylcarbethoxy)vinyl] phosphate, Pantozol 1, SD 4294, Volfazol, trans-Crotoxyphos |
| InChI formel | InChI=1S/C14H19O6P/c1-11(20-21(16,17-3)18-4)10-14(15)19-12(2)13-8-6-5-7-9-13/h5-10,12H,1-4H3/b11-10+ |
| SMIL | C(OC(/C=C(/OP(OC)(OC)=O)\C)=O)(C)C1=CC=CC=C1 |
| IUPAC navn | 1-phenylethyl (E)-3-dimethoxyphosphoryloxybut-2-enoate |
| Molekylvægt (g/mol) | 314.27 |
| Formel vægt | 314.0919 |