missing translation for 'onlineSavingsMsg'
Få mere at vide

trans-Clopidogrel-MP-13C,d3 Ethyl Ester Derivative(Mixture of Diastereomers), TRC

Artikelnummer. 30365520
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365520 5 mg
30367245 10 mg
30362875 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365520 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5872775MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1331383-23-3
Molekylær formel C2613CH27D3ClNO6S
Molekylvægt (g/mol) 536.06
Synonym Trans-Clopidogrel-Mp-13C,D3 Ethyl Ester Derivative, (Mixture Of Diastereomers)
IUPAC navn methyl 2-(2-chlorophenyl)-2-[(3E)-3-(2-ethoxy-2-oxoethylidene)-4-[2-oxo-2-[3-(trideuterio(113C)methoxy)phenyl]ethyl]sulfanylpiperidin-1-yl]acetate
SMIL [2H][13C]([2H])([2H])Oc1cccc(c1)C(=O)CSC2CCN(C/C/2=C\C(=O)OCC)C(C(=O)OC)c3ccccc3Cl

Tekniske data

Kemisk navn eller materiale trans-Clopidogrel-MP-13C,d3 Ethyl Ester Derivative(Mixture of Diastereomers)
Mængde 5 mg
InChI formel InChI=1S/C27H30ClNO6S/c1-4-35-25(31)15-19-16-29(26(27(32)34-3)21-10-5-6-11-22(21)28)13-12-24(19)36-17-23(30)18-8-7-9-20(14-18)33-2/h5-11,14-15,24,26H,4,12-13,16-17H2,1-3H3/b19-15+/i2+1D3
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.