missing translation for 'onlineSavingsMsg'
Få mere at vide

trans,cis-Sorbic acid, TRC

Artikelnummer. 30005829
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005829 25 mg
30004880 100 mg
30003679 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30005829 Leverandør Toronto Research Chemicals Leverandørnr. TRCS67689425MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 30361-30-9
Molekylær formel C6H8O2
Molekylvægt (g/mol) 112.13
Synonym (2E,4Z)-2,4-Hexadienoic acid, 2-trans,4-cis-Hexadienoic acid
IUPAC navn (2E,4Z)-hexa-2,4-dienoic acid
SMIL C\C=C/C=C/C(=O)O

Tekniske data

Kemisk navn eller materiale trans,cis-Sorbic acid
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2-,5-4+
Formel vægt 112.0524
Procent renhed >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.