Få mere at vide
trans,cis-Nepetalactone, TRC
Artikelnummer. 30480075
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480075 25 mg
30477482 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30480075 Leverandør Toronto Research Chemicals Leverandørnr. TRCN39005525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17257-15-7
Molekylær formel C10H14O2
Molekylvægt (g/mol) 166.22
Synonym [4aS-(4aα,7α,7aβ)]-5,6,7,7a-Tetrahydro-4,7-dimethyl-cyclopenta[c]pyran-1(4aH)-one, 5,6,7,7aβ-Tetrahydro-4,7α-dimethyl-cyclopenta[c]pyran-1(4aαH)-one, (4aS,7S,7aS)-5,6,7,7a-Tetrahydro-4,7-dimethylcyclopenta[c]pyran-1(4aH)-one, (+)-trans,cis-Nepetalactone, (4aS,7S,7aS)-Nepetalactone, 4aα,7α,7aβ-Nepetalactone, Epinepetalactone, Isonepetalactone
IUPAC navn (4aS,7S,7aS)-4,7-dimethyl-5,6,7,7a-tetrahydro-4aH-cyclopenta[c]pyran-1-one
SMIL CC(=O)NC1[C@@H](OC(CO)[C@H](O)C1O)Oc2cc3c(cc2)C(C)=CC(=O)O3
Tekniske data
Kemisk navn eller materiale trans,cis-Nepetalactone
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C10H14O2/c1-6-3-4-8-7(2)5-12-10(11)9(6)8/h5-6,8-9H,3-4H2,1-2H3/t6-,8+,9-/m0/s1
Formel vægt 166.0994
Procent renhed >95%