Få mere at vide
(±)-trans-Chrysanthemic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (±)-trans-Chrysanthemic Acid |
| CAS | 705-16-8 |
| Molekylær formel | C10H16O2 |
| Mængde | 1 g |
| Synonym | (1R,3R)-rel-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid (±)-trans-Chrysanthemumic Acid, (±)-trans-Chrysanthemummonocarboxylic Acid, DL-trans-Chrysanthemic Acid, dl-trans-Chrysanthemumic Acid, dl-trans-Chrysanthemummonocarboxylic Acid, trans-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid, trans-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid ester, trans-2,2-Dimethyl-3-(isobutenyl)cyclopropane-1-carboxylic Acid, trans-3,3-Dimethyl-2-(2-methyl-1-propenyl)cyclopropane-1-carboxylic Acid, trans-3,3-Dimethyl-2-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid, trans-Chrysanthemic Acid, trans-Chrysanthemum monocarboxylic Acid, trans-Chrysanthemumic Acid, (±)-trans-2,2-Dimethyl-3-isobutenyl-1-cyclopropanecarboxylic Acidtrans-(±)-2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylic Acid, trans-(±)-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic Acid |
| InChI formel | InChI=1S/C10H16O2/c1-6(2)5-7-8(9(11)12)10(7,3)4/h5,7-8H,1-4H3,(H,11,12)/t7-,8+/m1/s1 |
| SMIL | CC(=C[C@@H]1[C@@H](C(=O)O)C1(C)C)C |
| IUPAC navn | (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylic acid |
| Molekylvægt (g/mol) | 168.23 |
| Formel vægt | 168.115 |