Få mere at vide
trans-Carboxy Glimepiride-d5, TRC
Artikelnummer. 30349009
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30349009 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349009 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1784470.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-Carboxy Glimepiride-d5
CAS 1217718-14-3
Molekylær formel C24H27D5N4O7S
Mængde 0.5 mg
Synonym trans-4-[[[[[4-[2-[[(3-(Ethyl-d5)-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]amino]carbonyl]amino]cyclohexanecarboxylic Acid
InChI formel InChI=1S/C24H32N4O7S/c1-3-20-15(2)14-28(21(20)29)24(33)25-13-12-16-4-10-19(11-5-16)36(34,35)27-23(32)26-18-8-6-17(7-9-18)22(30)31/h4-5,10-11,17-18H,3,6-9,12-14H2,1-2H3,(H,25,33)(H,30,31)(H2,26,27,32)/i1D3,3D2
SMIL O=C(NS(C1=CC=C(CCNC(N2C(C(C([2H])([2H])C([2H])([2H])[2H])=C(C)C2)=O)=O)C=C1)(=O)=O)N[C@@H]3CC[C@@H](C(O)=O)CC3
IUPAC navn 4-[[4-[2-[[3-methyl-5-oxo-4-(1,1,2,2,2-pentadeuterioethyl)-2H-pyrrole-1-carbonyl]amino]ethyl]phenyl]sulfonylcarbamoylamino]cyclohexane-1-carboxylic acid
Molekylvægt (g/mol) 525.63