Få mere at vide
trans-Capsaicin-d3, TRC
Artikelnummer. 30331012
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30331012 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331012 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1756871MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-Capsaicin-d3
CAS 1217899-52-9
Renhedsgrad noter HPLC
Molekylær formel C18H24D3NO3
Mængde 1 mg
Synonym (6E)-N-[(4-hydroxy-3-(methoxy-d3)phenyl)methyl]-8-methyl-6-nonenamide, (E)-8-Methyl-N-vanillyl-6-nonenamide-d3, Capsaicin-d3, (E)-N-(4-Hydroxy-3- (methoxy-d3)benzyl)-8-methylnon-6-enamide, ALGRX 4975-d3, Adlea-d3, Axsain-d3, Capsaicine-d3, Capsin P 50-d3, Capzasin-HP-d3, Dolenon-d3, E-Capsaicin-d3, Mioton-d3, N-(4-Oxy-3-(methoxy-d3)benzyl)-8-methyl-6-nonenamide, NGX 4010-d3, NSC 56353-d3, Ovocap-d3, Ratden PE 40-d3, Togarashi Orenji-d3, Zostrix-d3, trans-8-Methyl-N-vanillyl-6-nonenamide-d3
InChI formel InChI=1S/C18H27NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,14,20H,4-5,7,9,13H2,1-3H3,(H,19,21)/b8-6+
SMIL O=C(CCCC/C=C/C(C)C)NCC1=CC(OC([2H])([2H])[2H])=C(O)C=C1
IUPAC navn (E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
Molekylvægt (g/mol) 308.43
Procent renhed >95