Få mere at vide
trans-Beta-Ocimene, TRC
Artikelnummer. 30012345
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012345 100 mg
30009629 50 mg
17960325 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012345 Leverandør Toronto Research Chemicals Leverandørnr. TRCO150025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3779-61-1
Molekylær formel C10 H16
Molekylvægt (g/mol) 136.23
Synonym 3E)-3,7-Dimethyl-1,3,6-octatriene, (3E)-Ocimene, (E)-Ocimene, (E)-β-Ocimene, trans-3,7-Dimethyl-1,3,6-octatriene, trans-β-Ocimene, β-trans-Ocimene, trans-β-Ocimene, trans-beta-Ocimene, trans-3,7-Dimethyl-1,3,6-octatriene
IUPAC navn (3E)-3,7-dimethylocta-1,3,6-triene
SMIL C/C(C=C)=C\C/C=C(C)/C
Tekniske data
Kemisk navn eller materiale trans-beta-Ocimene
Mængde 100 mg
InChI formel InChI=1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7-8H,1,6H2,2-4H3/b10-8+
Formel vægt 136.1252