Få mere at vide
(+)-trans-Allethrin (may contain up to 10% cis isomer), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | S-Bioallethrin |
| CAS | 28434-00-6 |
| Molekylær formel | C19 H26 O3 |
| Mængde | 10 mg |
| Synonym | Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, [5-(phenylmethyl)-3-furanyl]methyl ester, (1R,3R)- (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, [5-(phenylmethyl)-3-furanyl]methyl ester, (1R-trans)-, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, (5-benzyl-3-furyl)methyl ester, trans-(+)- (8CI), 3-Furanmethanol, 5-benzyl-, 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylate, trans-(+)- (8CI), (+)-trans-Bioresmethrin, (+)-trans-Resmethrin, 1R-trans-Resemethrin, 5-Benzyl-3-furylmethyl (+)-trans-chrysanthemate, AI 3-27662, Bioresmethrin, FMC 18739, Isathrine, Isatrin, NIA 18739, NRDC 107, Penick 1390, Pyrethroid NRDC 107, RU 11484, Resbuthrin, SBP 1390, d-trans-(5-Benzyl-3-furyl)methyl chrysanthemate, d-trans-Chrysron, d-trans-Resmethrin, d-trans-[(5-Benzyl-3-furyl)methyl] chrysanthemumate, (S)-Bioallethrin, S-Bioallethrin |
| InChI formel | InChI=1S/C19H26O3/c1-7-8-13-12(4)16(10-15(13)20)22-18(21)17-14(9-11(2)3)19(17,5)6/h7,9,14,16-17H,1,8,10H2,2-6H3/t14-,16+,17+/m1/s1 |
| SMIL | CC(=C[C@@H]1[C@@H](C(=O)O[C@H]2CC(=O)C(=C2C)CC=C)C1(C)C)C |
| IUPAC navn | [(1S)-2-methyl-4-oxo-3-prop-2-enylcyclopent-2-en-1-yl] (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate |
| Molekylvægt (g/mol) | 302.41 |
| Formel vægt | 302.1882 |