Få mere at vide
(±)-trans-9,10-Epoxystearic Acid Methyl Ester, TRC
Artikelnummer. 30371058
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30371058 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371058 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5899101G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-trans-9,10-Epoxystearic Acid Methyl Ester
CAS 6084-76-0
Molekylær formel C19 H36 O3
Mængde 1 g
Synonym (2R,3R)-rel-3-Octyl-2-oxiraneoctanoic Acid Methyl Ester, trans-9,10-Epoxy-octadecanoic Acid Methyl Ester, (2R,3R)-rel-3-Octyl-oxiraneoctanoic acid methyl ester, trans-3-Octyl-oxiraneoctanoic acid methyl ester, Methyl trans-9,10-epoxyoctadecanoate, Methyl trans-9,10-epoxystearate
InChI formel InChI=1S/C19H36O3/c1-3-4-5-6-8-11-14-17-18(22-17)15-12-9-7-10-13-16-19(20)21-2/h17-18H,3-16H2,1-2H3/t17-,18-/m0/s1
SMIL CCCCCCCC[C@@H]1O[C@H]1CCCCCCCC(=O)OC
IUPAC navn methyl 8-[(2S,3S)-3-octyloxiran-2-yl]octanoate
Molekylvægt (g/mol) 312.49
Formel vægt 312.2665