Learn More
Alfa Aesar™ trans-4-n-Butylcyclohexanecarboxylic acid, 99%
Brand: Alfa Aesar™ H57387.06
Additional Details : CAS Number : 38289-28-0 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
38289-28-0 | |
184.279 | |
BALGERHMIXFENA-UHFFFAOYSA-N | |
2060518 | |
CCCCC1CCC(CC1)C(=O)O |
C11H20O2 | |
MFCD00059560 | |
trans-4-butylcyclohexanecarboxylic acid, 4-butylcyclohexanecarboxylic acid, trans-4-n-butylcyclohexanecarboxylic acid, 4-butylcyclohexane carboxylic acid, 4-n-butylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-butyl, trans-4-butylcyclohexane carboxylic acid, 4-butylcyclohexanecarboxylic acid cis-and trans-mixture, cyclohexanecarboxylic acid, 4-butyl-, trans, enamine_005334 | |
4-butylcyclohexane-1-carboxylic acid |
Specifications
trans-4-n-Butylcyclohexanecarboxylic acid | |
38289-28-0 | |
MFCD00059560 | |
BALGERHMIXFENA-UHFFFAOYSA-N | |
4-butylcyclohexane-1-carboxylic acid | |
2060518 | |
99% |
5g | |
C11H20O2 | |
trans-4-butylcyclohexanecarboxylic acid, 4-butylcyclohexanecarboxylic acid, trans-4-n-butylcyclohexanecarboxylic acid, 4-butylcyclohexane carboxylic acid, 4-n-butylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-butyl, trans-4-butylcyclohexane carboxylic acid, 4-butylcyclohexanecarboxylic acid cis-and trans-mixture, cyclohexanecarboxylic acid, 4-butyl-, trans, enamine_005334 | |
CCCCC1CCC(CC1)C(=O)O | |
184.279 | |
184.28 |