missing translation for 'onlineSavingsMsg'
Få mere at vide

trans-4-Hydroxycyclohexanecarboxylic Acid, TRC

Artikelnummer. 30423282
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423282 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30423282 Leverandør Toronto Research Chemicals Leverandørnr. TRCR729038100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale trans-4-Hydroxycyclohexanecarboxylic Acid
CAS 3685-22-1
Molekylær formel C7H12O3
Mængde 100 mg
Synonym cis-4-Hydroxycyclohexanecarboxylic acid (ACI), cis-4-Hydroxycyclohexane-1-carboxylic acid
InChI formel InChI=1S/C7H12O3/c8-6-3-1-5(2-4-6)7(9)10/h5-6,8H,1-4H2,(H,9,10)/t5-,6+
SMIL O[C@@H]1CC[C@@H](CC1)C(=O)O
IUPAC navn 4-hydroxycyclohexane-1-carboxylic acid
Molekylvægt (g/mol) 144.17
Formel vægt 144.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.