missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals
301.57 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
trans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
trans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | ∼214°C (decomposition) |
Quantity | 25 g |
Beilstein | 2207383 |
Merck Index | 14,2560 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 164.16 |
Percent Purity | 98% |
Chemical Name or Material | trans-4-Hydroxycinnamic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'