Få mere at vide
trans-4-Hydroxy-2-hexenal-5,5,6,6,6-d5 Dimethyl Acetal, CDN
Artikelnummer. 30316637
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316637 0.01 g
30312648 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316637 Leverandør CDN Leverandørnr. CDND68210.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4 E)-6,6-Dimethoxy-4-hexen-3-ol-1,1,1,2,2-D5
CAS 1219806-40-2
Molekylær formel C8 D5 H11 O3
Mængde 0.01 g
Synonym (4 E)-6,6-Dimethoxy-4-hexen-3-ol-1,1,1,2,2-D5, trans-4-Hydroxy-2-hexenal-5,5,6,6,6-d5 dimethyl acetal
InChI formel InChI=1 S/C8H16O3/c1-4-7(9)5-6-8(10-2)11-3/h5-9 H,4H2,1-3H3/b6-5+/i1D3,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C(O)\C=C\C(OC)OC
IUPAC navn (E)-1,1,1,2,2-pentadeuterio-6,6-dimethoxyhex-4-en-3-ol
Molekylvægt (g/mol) 165.242
Formel vægt 165.141 g/mol
Procent renhed 99 atom % D, min 90% Chemical Purity